MAT184124078 Matrix Scientific

C-(4-Methyl-[1,2,3]thiadiazol-5-yl)-methylamine

  • MDL number MFCD10001570
  • Cas Number : 790641-72-4
  • Molecular Weight : 129.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/034672.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/034672.html
Availability details for
Quantity
  • Availability
  • Add to Cart