MAT184124124 Matrix Scientific

4-(1,1-Dioxo-1lambda*6*-[1,2]thiazinan-2-yl)-phenylamine

  • MDL number MFCD09261780
  • Cas Number : 37441-49-9
  • Molecular Weight : 226.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/034484.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/034484.html
Availability details for
Quantity
  • Availability
  • Add to Cart