MAT184124134 Matrix Scientific

(6-Amino-2,3-dihydro-indol-1-yl)-pyridin-3-yl-methanone

  • MDL number MFCD09261846
  • Cas Number : 927997-04-4
  • Molecular Weight : 239.28

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/034493.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/034493.html
Availability details for
Quantity
  • Availability
  • Add to Cart