MAT185013395 Matrix Scientific

4,6-Difluoropyrimidine

  • MDL number MFCD03412223
  • Cas Number : 2802-62-2
  • Molecular Weight : 116.07

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.matrixscientific.com/034743.html

Safety Information

hcodes Irritant/Refrigerate/Stored under Argon
MSDS http://www.matrixscientific.com/034743.html
Availability details for
Quantity
  • Availability
  • Add to Cart