MAT186689132 Matrix Scientific

1-Chloro-4-(1,1-difluoroethyl)-2,5-difluorobenzene

  • MDL number MFCD11226702
  • Cas Number : 1138444-89-9
  • Molecular Weight : 212.58

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Ambient
Country of Origin USA
Product Link http://www.matrixscientific.com/034774.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/034774.html
Availability details for
Quantity
  • Availability
  • Add to Cart