MAT187835291 Matrix Scientific

4-(4-Ethoxyphenyl)piperidine hydrochloride

  • MDL number MFCD11845766
  • Cas Number : 170856-82-3
  • Molecular Weight : 241.76

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/021743.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/021743.html
Availability details for
Quantity
  • Availability
  • Add to Cart