MAT187835311 Matrix Scientific

3-(1-Piperazino)propionitrile

  • MDL number MFCD02093566
  • Cas Number : 34064-86-3
  • Molecular Weight : 139.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Boiling point 95-98�m
Product Link http://www.matrixscientific.com/034753.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/034753.html
Availability details for
Quantity
  • Availability
  • Add to Cart