MAT188490643 Matrix Scientific

2-[(2-Bromoacetyl)amino]-N,N-dimethylbenzamide

  • MDL number MFCD12026550
  • Cas Number : 1138445-86-9
  • Molecular Weight : 285.15

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Storage temp. Ambient
Country of Origin USA
Product Link http://www.matrixscientific.com/034968.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/034968.html
Availability details for
Quantity
  • Availability
  • Add to Cart