MAT188490682 Matrix Scientific

2-[(2-Bromoacetyl)amino]-N-methylbenzamide

  • MDL number MFCD12026589
  • Cas Number : 1138442-44-0
  • Molecular Weight : 271.12

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/035009.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/035009.html
Availability details for
Quantity
  • Availability
  • Add to Cart