MAT188490727 Matrix Scientific

2-Bromo-N-[2-(diethylamino)ethyl]acetamide

  • MDL number MFCD12026637
  • Cas Number : 1138442-93-9
  • Molecular Weight : 237.15

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Storage temp. Freezer
Country of Origin USA
Product Link http://www.matrixscientific.com/035062.html

Safety Information

hcodes Irritant/Store in Freezer
MSDS http://www.matrixscientific.com/035062.html
Availability details for
Quantity
  • Availability
  • Add to Cart