MAT188490786 Matrix Scientific

1-Phenyl-1-propanamine hydrochloride

  • MDL number MFCD00267814
  • Cas Number : 24301-86-8
  • Molecular Weight : 171.67

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/034881.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/034881.html
Availability details for
Quantity
  • Availability
  • Add to Cart