MAT188490796 Matrix Scientific

1H-Indole-3-carbaldehyde oxime

  • MDL number MFCD00022737
  • Cas Number : 2592-05-4
  • Molecular Weight : 160.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 197-200
Country of Origin USA
Product Link http://www.matrixscientific.com/034889.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/034889.html
Availability details for
Quantity
  • Availability
  • Add to Cart