MAT188490804 Matrix Scientific

3-(3-Aminophenyl)-N,N-dimethylpropanamidehydrochloride

  • MDL number MFCD11226486
  • Cas Number : 1135288-85-5
  • Molecular Weight : 228.72

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/034896.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/034896.html
Availability details for
Quantity
  • Availability
  • Add to Cart