MAT188490837 Matrix Scientific

5-Oxo-5-[3-(1-piperidinylcarbonyl)anilino]-pentanoic acid

  • MDL number MFCD11226490
  • Cas Number : 1030497-05-2
  • Molecular Weight : 318.38

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/034921.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/034921.html
Availability details for
Quantity
  • Availability
  • Add to Cart