MAT189579602 Matrix Scientific

Methyl (2S,4S)-4-(2,5-difluorophenoxy)-2-pyrrolidinecarboxylate hydrochloride

  • MDL number MFCD12026511
  • Cas Number : 1217657-55-0
  • Molecular Weight : 293.7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/023704.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/023704.html
Availability details for
Quantity
  • Availability
  • Add to Cart