MAT189579640 Matrix Scientific

4-Amino-3-chloro-2,5,6-trifluoropyridine

  • MDL number MFCD00010856
  • Cas Number : 2693-57-4
  • Molecular Weight : 182.53

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 120-121
Purity 98
Country of Origin USA
Product Link http://www.matrixscientific.com/035279.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/035279.html
Availability details for
Quantity
  • Availability
  • Add to Cart