MAT196003978 Matrix Scientific

4-(Cyclohexylmethoxy)piperidine hydrochloride

  • MDL number MFCD12026488
  • Cas Number : 1185298-81-0
  • Molecular Weight : 233.78

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/014442.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/014442.html
Availability details for
Quantity
  • Availability
  • Add to Cart