MAT196004083 Matrix Scientific

{3-[3-(Morpholin-4-ylcarbonyl)phenyl]-1,2,4-oxadiazol-5-yl}methylamine hydrochloride

  • MDL number MFCD12026806
  • Cas Number : 1185299-40-4
  • Molecular Weight : 324.77

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/036209.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/036209.html
Availability details for
Quantity
  • Availability
  • Add to Cart