MAT196004114 Matrix Scientific

2-Carboxy-4-fluorophenylboronic acid

  • MDL number MFCD08235078
  • Cas Number : 874290-63-8
  • Molecular Weight : 183.93

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/035370.html

Safety Information

hcodes Irritant/Store in Freezer
MSDS http://www.matrixscientific.com/035370.html
Availability details for
Quantity
  • Availability
  • Add to Cart