MAT196004322 Matrix Scientific

3-(4-Chlorophenoxy)piperidine hydrochloride

  • MDL number MFCD04117749
  • Cas Number : 38247-51-7
  • Molecular Weight : 248.15

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/023204.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/023204.html
Availability details for
Quantity
  • Availability
  • Add to Cart