MAT196004519 Matrix Scientific

5-Bromo-2-methoxyphenol

  • MDL number MFCD00673137
  • Cas Number : 37942-01-1
  • Molecular Weight : 203.04

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin USA
Product Link http://www.matrixscientific.com/035780.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/035780.html
Availability details for
Quantity
  • Availability
  • Add to Cart