MAT196004537 Matrix Scientific

2-Ethyl-1,3-thiazole-4-carboxylic acid

  • MDL number MFCD09258842
  • Cas Number : 769124-05-2
  • Molecular Weight : 157.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/035798.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/035798.html
Availability details for
Quantity
  • Availability
  • Add to Cart