MAT196004607 Matrix Scientific

2-(6-Chloro-5-methyl-1,3-benzoxazol-2-yl)-ethanamine hydrochloride

  • MDL number MFCD11696375
  • Cas Number : 1158324-46-9
  • Molecular Weight : 247.13

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/035868.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/035868.html
Availability details for
Quantity
  • Availability
  • Add to Cart