MAT196004631 Matrix Scientific

1-(1,3-Benzoxazol-2-yl)piperidin-4-one

  • MDL number MFCD09701641
  • Cas Number : 1035840-42-6
  • Molecular Weight : 216.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/035892.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/035892.html
Availability details for
Quantity
  • Availability
  • Add to Cart