MAT196004690 Matrix Scientific

2-Amino-4-chloro-6-methylphenylamine

  • MDL number MFCD03426174
  • Cas Number : 109671-52-5
  • Molecular Weight : 156.62

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/035951.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/035951.html
Availability details for
Quantity
  • Availability
  • Add to Cart