MAT196004735 Matrix Scientific

1,3-Dimethyl-1H-pyrazole-4-carbaldehyde oxime

  • MDL number MFCD02006255
  • Cas Number : 380583-07-3
  • Molecular Weight : 139.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/035996.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/035996.html
Availability details for
Quantity
  • Availability
  • Add to Cart