MAT196004738 Matrix Scientific

2-(Pyridin-2-ylthio)acetohydrazide

  • MDL number MFCD01422111
  • Cas Number : 104496-91-5
  • Molecular Weight : 183.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/035999.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/035999.html
Availability details for
Quantity
  • Availability
  • Add to Cart