MAT196004748 Matrix Scientific

2-(Chloromethyl)-5-methyl-1,3-benzoxazole

  • MDL number MFCD07366537
  • Cas Number : 41014-44-2
  • Molecular Weight : 181.62

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/036009.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/036009.html
Availability details for
Quantity
  • Availability
  • Add to Cart