MAT196004776 Matrix Scientific

N-1-Adamantylhydrazinecarboxamide

  • MDL number MFCD00197260
  • Cas Number : 26496-36-6
  • Molecular Weight : 209.29

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/036037.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/036037.html
Availability details for
Quantity
  • Availability
  • Add to Cart