MAT196004789 Matrix Scientific

2-[(5-{1-[(3-Methylphenyl)amino]ethyl}-4-phenyl-4H-1,2,4-triazol-3-yl)thio]acetohydrazide

  • MDL number MFCD11696430
  • Cas Number : 1071368-77-8
  • Molecular Weight : 382.49

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/036050.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/036050.html
Availability details for
Quantity
  • Availability
  • Add to Cart