MAT196004808 Matrix Scientific

6-Chloro-2-hydrazino-1H-benzimidazole

  • MDL number MFCD11053045
  • Cas Number : 99122-11-9
  • Molecular Weight : 182.61

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/036069.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/036069.html
Availability details for
Quantity
  • Availability
  • Add to Cart