MAT196004851 Matrix Scientific

2-Methyl-3-pyrrolidin-1-ylpropan-1-ol

  • MDL number MFCD08061224
  • Cas Number : 873376-29-5
  • Molecular Weight : 143.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/036112.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/036112.html
Availability details for
Quantity
  • Availability
  • Add to Cart