MAT196004888 Matrix Scientific

2,5-Dichloro-1,3-benzoxazole

  • MDL number MFCD09264312
  • Cas Number : 3621-81-6
  • Molecular Weight : 188.01

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/036149.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/036149.html
Availability details for
Quantity
  • Availability
  • Add to Cart