MAT196004929 Matrix Scientific

[5-(4-Methoxyphenyl)-1,2,4-oxadiazol-3-yl]-methylamine, hydrochloride

  • Cas Number : 914810-86-9
  • Molecular Weight : 241.68

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/036190.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/036190.html
Availability details for
Quantity
  • Availability
  • Add to Cart