MAT196005029 Matrix Scientific

2-(Piperazin-1-ylsulfonyl)ethanamine oxalate

  • MDL number MFCD12027067
  • Cas Number : 1185299-71-1
  • Molecular Weight : 283.31

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/037168.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/037168.html
Availability details for
Quantity
  • Availability
  • Add to Cart