MAT196005054 Matrix Scientific

2-(Dimethylamino)butanoic acid

  • MDL number MFCD10686586
  • Cas Number : 170941-86-3
  • Molecular Weight : 131.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/036283.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/036283.html
Availability details for
Quantity
  • Availability
  • Add to Cart