MAT196005076 Matrix Scientific

2-(4-Methylphenyl)-2-piperidin-1-ylethanamine

  • MDL number MFCD08667757
  • Cas Number : 927965-82-0
  • Molecular Weight : 218.34

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/036305.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/036305.html
Availability details for
Quantity
  • Availability
  • Add to Cart