MAT196005165 Matrix Scientific

4-[(4-Methoxyphenoxy)methyl]-5-methylisoxazole-3-carboxylic acid

  • MDL number MFCD06805578
  • Cas Number : 932918-29-1
  • Molecular Weight : 263.25

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/036394.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/036394.html
Availability details for
Quantity
  • Availability
  • Add to Cart