MAT196005253 Matrix Scientific

4-Methoxy-3-[(3-methylpiperidin-1-yl)methyl]-benzaldehyde

  • MDL number MFCD12026917
  • Cas Number : 1119449-75-0
  • Molecular Weight : 247.34

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/036482.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/036482.html
Availability details for
Quantity
  • Availability
  • Add to Cart