MAT196005286 Matrix Scientific

4,5-Dimethylisoxazole-3-carboxylic acid

  • MDL number MFCD05667138
  • Cas Number : 100047-61-8
  • Molecular Weight : 141.13

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/036515.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/036515.html
Availability details for
Quantity
  • Availability
  • Add to Cart