MAT196005358 Matrix Scientific

3-(4-Bromo-3,5-dimethyl-1H-pyrazol-1-yl)-propanenitrile

  • MDL number MFCD00475560
  • Cas Number : 90006-21-6
  • Molecular Weight : 228.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/036587.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/036587.html
Availability details for
Quantity
  • Availability
  • Add to Cart