MAT196005554 Matrix Scientific

(4-Methoxy-3,5-dimethylpyridin-2-yl)methylaminedihydrochloride

  • MDL number MFCD12026975
  • Cas Number : 130000-78-1
  • Molecular Weight : 239.15

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/036783.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/036783.html
Availability details for
Quantity
  • Availability
  • Add to Cart