MAT196005555 Matrix Scientific

1-(Cyclopropylcarbonyl)piperidin-3-amine

  • MDL number MFCD12026976
  • Cas Number : 1114596-39-2
  • Molecular Weight : 168.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/036784.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/036784.html
Availability details for
Quantity
  • Availability
  • Add to Cart