MAT196005582 Matrix Scientific

1-[(4-Methoxyphenyl)sulfonyl]piperidin-4-amine

  • MDL number MFCD06016003
  • Cas Number : 728015-65-4
  • Molecular Weight : 270.35

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/036811.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/036811.html
Availability details for
Quantity
  • Availability
  • Add to Cart