MAT196005589 Matrix Scientific

1-Methyl-1H-indole-4-carbonitrile

  • MDL number MFCD08690265
  • Cas Number : 628711-58-0
  • Molecular Weight : 156.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/036818.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/036818.html
Availability details for
Quantity
  • Availability
  • Add to Cart