MAT196005599 Matrix Scientific

2-(Chloromethyl)-3,5-dimethylpyridin-4(1H)-onehydrochloride

  • MDL number MFCD12026993
  • Cas Number : 220770-99-0
  • Molecular Weight : 208.09

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Storage temp. Ambient
Country of Origin USA
Product Link http://www.matrixscientific.com/036828.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/036828.html
Availability details for
Quantity
  • Availability
  • Add to Cart