MAT196005603 Matrix Scientific

2-Amino-N-[2-(5-chloro-1H-indol-3-yl)ethyl]-acetamidehydrochloride [Stabilized with 15% dioxane]

  • MDL number MFCD12026996
  • Cas Number : 1158282-91-7
  • Molecular Weight : 288.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/036832.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/036832.html
Availability details for
Quantity
  • Availability
  • Add to Cart