MAT196005618 Matrix Scientific

1,4-Dimethyl-1H-indole-3-carbaldehyde

  • MDL number MFCD11505328
  • Cas Number : 170489-16-4
  • Molecular Weight : 173.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/036847.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/036847.html
Availability details for
Quantity
  • Availability
  • Add to Cart