MAT196005664 Matrix Scientific

1-(Methylsulfonyl)-4,4'-bipiperidine

  • MDL number MFCD06740340
  • Cas Number : 263393-49-3
  • Molecular Weight : 246.37

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/036893.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/036893.html
Availability details for
Quantity
  • Availability
  • Add to Cart