MAT196005669 Matrix Scientific

1-[2-(4-Methoxyphenyl)ethyl]piperazine

  • MDL number MFCD07365315
  • Cas Number : 55455-94-2
  • Molecular Weight : 220.32

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/036898.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/036898.html
Availability details for
Quantity
  • Availability
  • Add to Cart